Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Antibody type:Primary
- Antigen name:mucin 6, oligomeric mucus/gel-forming
- Antigen symbol:MUC6
- Clonality:Monoclonal
- Clone:MUCN6-1
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG1 kappa
- Reactivity:Human
- Format:Purified Antibody
- Form:Liquid
- Gene ID:Q6W4X9
- Antigen synonyms:MUC6
- Storage buffer:PBS with 0,1 mg/ml BSA and 0,05% sodium azide
- Storage temperature:Aliquot and Store at 2…8 °C. Avoid freez-thaw cycles.
- Concentration:0,2 mg/ml
- Shipping temperature:4 °C
- Immunogen:Recombinant human protein was used as the immunogen for the Mucin-6 antibody.
- Purification:Protein G affinity chromatography
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Mucin 6 may provide a mechanism for modulation of the composition of the protective mucus layer related to acid secretion or the presence of bacteria and noxious agents in the lumen. Plays an important role in the cytoprotection of epithelial surfaces and are used as tumor markers in a variety of cancers. May play a role in epithelial organogenesis. Highest expression seen in the stomach and gall bladder, with lower expression in the terminal ileum and right colon. [UniProt].
Immunofluorescence: 1 - 2 µg/ml Immunohistochemistry (FFPE): 2-4 µg/ml for 30 min at RT (1) Prediluted format: incubate for 30 min at RT (2) Titering of the Mucin-6 antibody may be required for optimal performance.
1. FFPE testing requires sections to be boiled in pH6 10 mM citrate buffer for 10 - 20 minutes, followed by cooling at RT for 20 minutes, prior to staining.
2. The prediluted format is supplied in a dropper bottle and is optimised for use in IHC. After epitope retrieval step (if required), drip mAb solution onto the tissue section and incubate at RT for 30 min.
Type: Primary
Antigen: MUC6
Clonality: Monoclonal
Clone: MUCN6-1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1, kappa
Reactivity: Human