Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
- Antibody type:Primary
- Antigen name:mucin 2, oligomeric mucus/gel-forming
- Antigen symbol:MUC2
- Clonality:Monoclonal
- Clone:MUCN2-1
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG1 kappa
- Format:Purified Antibody
- Form:Liquid
- Gene ID:Q02817
- Antigen synonyms:MUC2
- Storage buffer:PBS with 0,1 mg/ml BSA and 0,05% sodium azide
- Storage temperature:Aliquot and Store at 2…8 °C. Avoid freez-thaw cycles.
- Concentration:0,2 mg/ml
- Shipping temperature:4 °C
- Immunogen:Recombinant human protein was used as the immunogen for the MUC2 antibody.
- Purification:Protein G affinity chromatography
- Size:100 µg
- Pk:100 µG
Mucin 2 coats the epithelia of the intestines, airways, and other mucus membrane-containing organs. Thought to provide a protective, lubricating barrier against particles and infectious agents at mucosal surfaces. Major constituent of both the inner and outer mucus layers of the colon and may play a role in excluding bacteria from the inner mucus layer. [UniProt].
Immunofluorescence: 1 - 2 µg/ml Immunohistochemistry (FFPE): 1 - 2 µg/ml for 30 min at RT (1) Prediluted format: incubate for 30 min at RT (2) Titering of the MUC2 antibody may be required for optimal performance.
1. Staining of formalin-fixed tissues requires boiling tissue sections in 10 mM Tris with 1mM EDTA, pH 9.0, for 10 - 20 min followed by cooling at RT for 20 minutes
2. The prediluted format is supplied in a dropper bottle and is optimised for use in IHC. After epitope retrieval step (if required), drip mAb solution onto the tissue section and incubate at RT for 30 min.
Type: Primary
Antigen: MUC2
Clonality: Monoclonal
Clone: MUCN2-1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1, kappa
Reactivity: Human