Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Antibody type:Primary
- Antigen name:spectrin beta, non-erythrocytic 2
- Antigen symbol:SPTBN2
- Clonality:Monoclonal
- Clone:SPTBN2/1582
- Conjugation:Unconjugated
- ELISA:Yes
- Flow cytometry:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG2a kappa
- Western blot:Yes
- Format:Purified Antibody
- Form:Liquid
- Gene ID:O15020
- Antigen synonyms:SPTBN2
- Storage buffer:PBS with 0,1 mg/ml BSA and 0,05% sodium azide
- Storage temperature:Aliquot and Store at 2…8 °C. Avoid freez-thaw cycles.
- Concentration:0,2 mg/ml
- Shipping temperature:4 °C
- Immunogen:Human recombinant partial protein corresponding to amino acids 356-475 was used as the immunogen for this Spectrin beta III antibody.
- Purification:Protein G affinity chromatography
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Spectrin is an actin binding protein that is a major component of the plasma membrane skeleton. Spectrins function as membrane organizers and stabilizers by forming dimers, tetramers and higher polymers. Spectrin alpha and spectrin beta are present in erythrocytes, whereas spectrin alphaII (fodrin alpha) and spectrin betaI (fodrin beta) are present in other somatic cells. SPTBN2/Spectrin betaIII is highly homologous to both spectrin betaI and spectrin betaII. SPTBN2 is highly expressed in brain, kidney, pancreas and liver, and at lower levels in lung and placenta.
Flow Cytometry: 0,5 - 1 µg/10^6 cells in 0,1 ml IF: 1 - 2 µg/ml IHC (FFPE): 0,5 - 1 µg/ml for 30 min at RT WB: 0,5 - 1 µg/ml ELISA: 2-4 µg/ml; order the BSA-free formulation for coating The concentration stated for each application is a general starting point. Variations in protocols, secondaries and substrates may require the Spectrin beta III antibody to be titered up or down for optimal performance.
Type: Primary
Antigen: SPTBN2
Clonality: Monoclonal
Clone: SPTBN2/1582
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a, kappa
Reactivity: Human