Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
- Antibody type:Primary
- Antigen name:membrane-spanning 4-domains, subfamily A, member 1
- Antigen symbol:MS4A1
- Clonality:Monoclonal
- Clone:93-1B3
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Host:Mouse
- ImmunoFluorescence:Yes
- Isotype:IgG1 kappa
- Western blot:Yes
- Neutralisation:Yes
- Format:Purified Antibody
- Form:Liquid
- Gene ID:P11836
- Antigen synonyms:MS4A1
- Storage buffer:PBS with 0,1 mg/ml BSA and 0,05% sodium azide
- Storage temperature:Aliquot and Store at 2…8 °C. Avoid freez-thaw cycles.
- Concentration:0,2 mg/ml
- Shipping temperature:4 °C
- Immunogen:Stimulated human leukocytes were used as the immunogen for this CD20 antibody.
- Tested applications:Functional
- Purification:Protein G affinity chromatography
- Size:100 µg
- Pk:100 µG
This antibody recognises a protein of 33-37 kDa, identified as CD20 (Workshop V; Code CD20.12). The antibody recognises the extracellular domain of the protein. The epitope is similar to or identical to that recognized by other CD20 antibodies including Leu-16 and B1. This antibody can be used for immunophenotyping of leukemia and malignant cells, B lymphocyte detection in peripheral blood, Bcell localization in tissues and B lymphocyte purification by immunosorbent methods. CD20 is a non-Ig differentiation antigen of Bcells and its expression is restricted to normal and neoplastic Bcells, being absent from all other leukocytes and tissues. It is expressed by pre Bcells and persists during all stages of Bcell maturation but is lost upon terminal differentiation into plasma cells. Protein passes through the membrane 4 times with both ends in cytoplasm and exposes one short and one longer loop to the external environment. CD20 is not glycosylated in resting Bcells and its cytoplasmic domains are differentially phosphorylated upon activation. It acts as a calcium channel involved in Bcell activation and cell cycle progression.
Flow Cytometry: 0,5-1 µg/million cells IF: 0,5 - 1 µg/ml Functional Testing (Order Ab without BSA & Azide) The concentration stated for each application is a general starting point. Variations in protocols, secondaries and substrates may require the CD20 antibody to be titered up or down for optimal performance.
Type: Primary
Antigen: MS4A1
Clonality: Monoclonal
Clone: 93-1B3
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1, kappa
Reactivity: Human