Order Entry
Northern Ireland
ContactUsLinkComponent
Anti-XRCC6 Mouse Monoclonal Antibody [clone: KU729]
Anti-XRCC6 Mouse Monoclonal Antibody [clone: KU729]
Catalog # PRSI33-138
Supplier:  ProSci Inc.
Anti-XRCC6 Mouse Monoclonal Antibody [clone: KU729]
Catalog # PRSI33-138
Supplier:  ProSci Inc.

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    X-ray repair complementing defective repair in Chinese hamster cells 6
  • Antigen symbol:
    XRCC6
  • Clonality:
    Monoclonal
  • Clone:
    KU729
  • Conjugation:
    Unconjugated
  • Flow cytometry:
    Yes
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG1 kappa
  • Format:
    Purified Antibody
  • Form:
    Liquid
  • Gene ID:
    Q6FG89
  • Antigen synonyms:
    XRCC6
  • Storage buffer:
    PBS with 0,1 mg/ml BSA and 0,05% sodium azide
  • Storage temperature:
    Aliquot and Store at 2…8 °C. Avoid freez-thaw cycles.
  • Concentration:
    0,2 mg/ml
  • Shipping temperature:
    4 °C
  • Immunogen:
    Nuclear extract of human HL-60 cells was used as the immunogen for this Ku70 + Ku80 antibody.
  • Tested applications:
    Immunocytochemistry
  • Purification:
    Protein G purified monoclonal antibody
  • Size:
    100 µg
  • Pk:
    100 µG

Specifications

About this item

This antibody recognises a dimer of two proteins of 70 kDa (Ku70) and ~80 kDa (Ku80), identified as two subunits of Ku. Antbody KU729 recognises a conformational epitope of the Ku70 + Ku80 dimer, which is destroyed during Western blotting. The Ku70 + Ku80 dimer is important for function of a 460 kDa DNA-dependent protein kinase. Ku protein plays a role in cell signaling, proliferation, DNA repair, replication, transcriptional activation, and apoptosis.

Flow cytometry: 0,5 - 1 µg/10e6 cells IF: 0,5-1,0 µg/ml Immunocytochemistry (Acetone-fixed cells): 0,25-0,5 µg/ml for 30 minutes at RT The concentration stated for each application is a general starting point. Variations in protocols, secondaries and substrates may require the Ku70 + Ku80 antibody to be titered up or down for optimal performance.

Type: Primary
Antigen: XRCC6
Clonality: Monoclonal
Clone: KU729
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1, kappa
Reactivity: Human, Primates