Order Entry
Northern Ireland
ContactUsLinkComponent
Anti-ACSS2 Rabbit Polyclonal Antibody
Anti-ACSS2 Rabbit Polyclonal Antibody
Catalog # PRSI15-095
Supplier:  ProSci Inc.
Anti-ACSS2 Rabbit Polyclonal Antibody
Catalog # PRSI15-095
Supplier:  ProSci Inc.

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Acetyl-coenzyme A synthetase, cytoplasmic
  • Antigen symbol:
    ACSS2
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG
  • Western blot:
    Yes
  • Form:
    Liquid
  • Gene ID:
    55902
  • Antigen synonyms:
    AceCS|Acetyl-CoA synthetase|Acetate--CoA ligase|ACS|Acyl-CoA synthetase short-chain family member 2|cytoplasmic|ACAS2|Acetyl-coenzyme A synthetase|Acyl-activating enzyme|ACSS2
  • Modification:
    None
  • Storage buffer:
    PBS with 0,02% sodium azide, 50% glycerol, pH 7,3
  • Storage temperature:
    Store at −20 °C. Avoid freeze/thaw cycles
  • Concentration:
    Batch dependent
  • Shipping temperature:
    20
  • Immunogen:
    Recombinant fusion protein containing a sequence corresponding to amino acids 367-606 of human ACSS2 (NP_001229322.1)
  • Purification:
    Affinity purification
  • Pack type:
    Vial
  • Pk:
    50 µl

Specifications

About this item

Type: Primary
Antigen: ACSS2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat