Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Antibody type:Primary
- Antigen name:Integrin alpha M
- Antigen symbol:Itgam
- Clonality:Monoclonal
- Clone:[M1/70]
- Conjugation:APC (Allophycocyanin)-Cy7®
- Flow cytometry:Yes
- Host:Rat
- Isotype:IgG2b
- Reactivity:Baboon,Rabbit,Rhesus monkey,Human,Mouse
- Form:Liquid
- Gene ID:16409
- Antigen synonyms:C3biR|Mo1|CR3|Mac-1|Mac-1alpha|Integrin alphaM
- Modification:None
- Storage buffer:Supplied in PBS/NaN₃ and a stabilising agent
- Storage temperature:Store vial at 2...8 °C
- Concentration:0,1 mg/ml
- Shipping temperature:20
- Immunogen:C57BL/10 mouse splenic T cells and concanavalin A-activated C57BL/10 splenocytes
- Pack type:Vial
- Pk:0,1 mg
Specifications
About this item
Type: Primary
Antigen: Itgam
Clonality: Monoclonal
Clone: [M1/70]
Conjugation: APC (Allophycocyanin)-Cy7®
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity: Mouse, Human, Rhesus Monkey, Baboon, Rabbit