Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Antibody type:Primary
- Antigen name:Acetylserotonin O-methyltransferase
- Antigen symbol:ASMT
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Form:Liquid
- Gene ID:438
- Antigen synonyms:HIOMT|ASMT|Acetylserotonin O-methyltransferase|Hydroxyindole O-methyltransferase
- Modification:None
- Storage buffer:PBS with 0,02% sodium azide, pH 7,3
- Storage temperature:Store at 4...730 °C. Avoid freeze/thaw cycles.
- Concentration:Batch dependent
- Shipping temperature:20
- Immunogen:A synthetic peptide of human ASMT
- Purification:Affinity purification
- Pack type:Vial
- Pk:100 µl
Specifications
About this item
Type: Primary
Antigen: ASMT
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human