Order Entry
Northern Ireland
ContactUsLinkComponent
Anti-CD4 Mouse Monoclonal Antibody [clone: RPA-T4] (PB (Pacific Blue))
Anti-CD4 Mouse Monoclonal Antibody [clone: RPA-T4] (PB (Pacific Blue))
Supplier:  AAT BIOQUEST
Certificates
undefined
Anti-CD4 Mouse Monoclonal Antibody [clone: RPA-T4] (PB (Pacific Blue))
Supplier:  AAT BIOQUEST

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Catalog No:
  • AATB100411J0
  • AATB100411J1
  • Antigen symbol:
  • CD4
  • CD4
  • Antigen name:
  • CD4 antigen
  • CD4 antigen
  • Antibody type:
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • PB (Pacific Blue)
  • PB (Pacific Blue)
  • Clone:
  • RPA-T4
  • RPA-T4
  • Reactivity:
  • Human
  • Human
  • Host:
  • Mouse
  • Mouse
  • Isotype:
  • IgG1
  • IgG1
  • ImmunoFluorescence:
  • Yes
  • Yes
  • Flow cytometry:
  • Yes
  • Yes
  • Purification:
  • Affinity chromatography
  • Affinity chromatography
  • Cross adsorption:
  • No
  • No
  • Storage temperature:
  • 2...8 °C with minimised light exposure. Do not freeze.
  • 2...8 °C with minimised light exposure. Do not freeze.
  • Shipping temperature:
  • 20
  • 20

Specifications

Frequently Bought Together