Order Entry
Northern Ireland
ContactUsLinkComponent
Eye Lysate (7 Days Old)
Eye Lysate (7 Days Old)
Catalog # PRSI1420-7
Supplier:  ProSci Inc.
CAS Number:  
Eye Lysate (7 Days Old)
Catalog # PRSI1420-7
Supplier:  ProSci Inc.
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Cell Format:
    Cryopreserved
  • Pk:
    0,1 mg
  • Description:
    Normal; Source: Mouse; Tissue: Eye
  • Storage conditions:
    Store at 2...8 °C for continuous use. For extended storage, freeze working aliquots at −70 °C.
  • Storage buffer:
    Eye lysate (7 Days Old) is supplied in SDS sample buffer containing 5% β-mercaptoethanol
  • Concentration:
    2 mg/ml
  • Tested applications:
    WB

Specifications

About this item

Eye tissue lysate (7 Days Old) was prepared by homogenisation in modified RIPA buffer (150 mM sodium chloride, 50 mM Tris-HCl, pH 7,4, 1 mM ethylenediaminetetraacetic acid, 1 mM phenylmethylsulfonyl fluoride, 1% Triton X-100, 1% sodium deoxycholic acid, 0,1% sodium dodecylsulfate, 5 μg/ml of aprotinin, 5 μg/ml of leupeptin.

Tissue and cell debris was removed by centrifugation. Protein concentration was determined with Bio-Rad protein assay. The product was boiled for 5 min in 1 x SDS sample buffer (50 mM Tris-HCl pH 6,8, 12,5% glycerol, 1% sodium dodecylsulfate, 0,01% bromophenol blue) containing 5% β-mercaptoethanol.