Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Antibody type:Primary
- Antigen name:Insulin-like growth factor 2 mRNA-binding protein 2
- Antigen symbol:IGF2BP2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human
- Western blot:Yes
- Epitope:Amino Acid Range = 530 - 556
- Form:Liquid
- Gene ID:Q9Y6M1
- Antigen synonyms:IGF2BP2
- Modification:None
- Storage buffer:Supplied in PBS with 0,09% (W/V) sodium azide.
- Molecular weight:66 kDa
- Storage temperature:Store at 4 °C for three months and –20 °C, stable for up to one year. As with all antibodies care should be taken to avoid repeated freeze thaw cycles. Antibodies should not be exposed to prolonged high temperatures.
- Shipping temperature:4 °C
- Immunogen:This IGF2BP2 antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 530 - 556 amino acids from the C-terminal region of human IGF2BP2
- Purification:This antibody is purified through a protein A column, followed by peptide affinity purification.
- Pk:400 µl
Specifications
About this item
This gene encodes a member of the IGF-II mRNA-binding protein (IMP) family. The protein encoded by this gene contains several four KH domains and two RRM domains. It functions by binding to the 5' UTR of the insulin-like growth factor 2 (IGF2) mRNA and regulating IGF2 translation. Alternate transcriptional splice variants, encoding different isoforms, have been characterized.
For WB starting dilution is: 1:1000. For IHC-P starting dilution is: 1:50~100. For FACS starting dilution is: 1:10~50.
Type: Primary
Antigen: IGF2BP2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Amino Acid Range = 530-556
Host: Rabbit
Isotype:
Reactivity: Human