Order Entry
Northern Ireland
ContactUsLinkComponent
Anti-Ptprc Rat Monoclonal Antibody [clone: 30-F11] (APC (Allophycocyanin))
Anti-Ptprc Rat Monoclonal Antibody [clone: 30-F11] (APC (Allophycocyanin))
Catalog # PRSI76-534
Supplier:  ProSci Inc.
Anti-Ptprc Rat Monoclonal Antibody [clone: 30-F11] (APC (Allophycocyanin))
Catalog # PRSI76-534
Supplier:  ProSci Inc.

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    protein tyrosine phosphatase, receptor type, C
  • Antigen symbol:
    Ptprc
  • Clonality:
    Monoclonal
  • Clone:
    30-F11
  • Conjugation:
    APC (Allophycocyanin)
  • Flow cytometry:
    Yes
  • Host:
    Rat
  • Isotype:
    IgG2b kappa
  • Cross adsorption:
    No
  • Form:
    liquid
  • Gene ID:
    P06800
  • Antigen synonyms:
    Ptprc
  • Storage buffer:
    Phosphate-buffered aqueous solution, ≤0,09% Sodium azide, may contain carrier protein/stabiliser, ph 7,2.
  • Storage temperature:
    The product should be stored undiluted at 4 °C and should be protected from prolonged exposure to light. Do not freeze.
  • Shipping temperature:
    4 °C
  • Purification:
    The monoclonal antibody was purified utilizing affinity chromatography and unreacted dye was removed from the product.
  • Pk:
    0,025 mg

Specifications

About this item

Type: Primary
Antigen: Ptprc
Clonality: Monoclonal
Clone: 30-F11
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Rat
Isotype: IgG2b kappa
Reactivity: Mouse