Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Antibody type:Primary
- Antigen name:Transcription factor PU.1
- Antigen symbol:SPI1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Reactivity:Human
- Western blot:Yes
- Epitope:217 - 246 aa
- Form:Liquid
- Gene ID:6688
- Modification:None
- Molecular weight:31 kDa
- Storage temperature:Store at 4 °C for three months and –20 °C, stable for up to one year. As with all antibodies care should be taken to avoid repeated freeze thaw cycles. Antibodies should not be exposed to prolonged high temperatures.
- Concentration:0,45 mg/ml
- Shipping temperature:4 °C
- Immunogen:This SPI1 antibody is generated from rabbits immunised with a KLH conjugated synthetic peptide between 217-246 amino acids from the C-terminal region of human SPI1
- Purification:This antibody is purified through a protein A column, followed by peptide affinity purification
- Pk:400 µl
Specifications
About this item
For IF starting dilution is: 1:10~50. For WB starting dilution is: 1:1000. For IHC-P starting dilution is: 1:10~50.
Type: Primary
Antigen: SPI1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Amino Acid Range = 217-246
Host: Rabbit
Isotype: Ig
Reactivity: Human