Order Entry
Northern Ireland
ContactUsLinkComponent
Anti-BAY 1112623 Recombinant Antibody
Anti-BAY 1112623 Recombinant Antibody
Catalog # PRSI10-781
Supplier:  ProSci Inc.
New Product
Anti-BAY 1112623 Recombinant Antibody
Catalog # PRSI10-781
Supplier:  ProSci Inc.
New Product
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Antibody type:
    Primary
  • Clonality:
    Recombinant
  • Conjugation:
    Unconjugated
  • Host:
  • Isotype:
    IgG4 kappa
  • Reactivity:
  • Environmentally Preferable:
  • Antigen synonyms:
    UCB-7665
  • Modification:
    None
  • Storage buffer:
    PBS buffer pH 7,5
  • Storage temperature:
    Use a manual defrost freezer and avoid repeated freeze thaw cycles. Store at 2…8 °C for 1 - 2 weeks. Store at –20 °C for 12 months. Store at –80 °C for long term storage.
  • Concentration:
    Batch dependent
  • Shipping temperature:
    20
  • Immunogen:
    Chimeric,Humanised/FCGRT [Homo sapiens]
  • Purification:
    >95%
  • Pack type:
    Vial
  • Pk:
    0,1 mg

Specifications

About this item

Type: Primary
Antigen:
Clonality: Recombinant
Clone:
Conjugation: Unconjugated
Epitope:
Host:
Isotype: IgG4-kappa
Reactivity: