Order Entry
Northern Ireland
ContactUsLinkComponent
Anti-CETP Rabbit Polyclonal Antibody
Anti-CETP Rabbit Polyclonal Antibody
Catalog # PRSI62-912
Supplier:  ProSci Inc.
undefined
Anti-CETP Rabbit Polyclonal Antibody
Catalog # PRSI62-912
Supplier:  ProSci Inc.

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Cholesteryl ester transfer protein {ECO:0000303|PubMed:3600759}
  • Antigen symbol:
    CETP
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Epitope:
    34-65 aa
  • Form:
    Liquid
  • Gene ID:
    1071
  • Modification:
    None
  • Molecular weight:
    55 kDa
  • Storage temperature:
    Store at 4 °C for three months and –20 °C, stable for up to one year. As with all antibodies care should be taken to avoid repeated freeze thaw cycles. Antibodies should not be exposed to prolonged high temperatures.
  • Concentration:
    2 mg/ml
  • Shipping temperature:
    4 °C
  • Immunogen:
    This CETP antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 34-65 amino acids from the N-terminal region of human CETP.
  • Purification:
    This antibody is prepared by Saturated Ammonium Sulfate (SAS) precipitation followed by dialysis
  • Pk:
    400 µl

Specifications

About this item

For WB starting dilution is: 1:1000.

Type: Primary
Antigen: CETP
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Amino Acid Range = 34-65
Host: Rabbit
Isotype: Ig
Reactivity: Human