Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
- Antibody type:Primary
- Antigen name:C-C motif chemokine receptor 5
- Antigen symbol:CCR5
- Clonality:Recombinant
- Clone:RC-10
- Conjugation:Unconjugated
- ELISA:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG kappa
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Form:Liquid
- Gene ID:1234
- Antigen synonyms:CD195|C-C CKR-5|C-C chemokine receptor type 5|CC-CKR-5|CHEMR13|CCR-5|HIV-1 fusion coreceptor
- Modification:Unmodified
- Storage buffer:PBS with 0,02% Proclin 300
- Storage temperature:Store at 4 °C for up to 3 months. For longer storage, aliquot and store at −20 °C.
- Shipping temperature:Shipped on blue ice
- Immunogen:This antibody was raised by immunizing mice with CCR5
- Pack type:Vial
- Pk:0,2 mg
Type: Primary
Antigen: C-C motif chemokine receptor 5
Clonality: Recombinant
Clone: RC-10
Conjugation: Unconjugated
Epitope:
Host:
Isotype: IgG kappa
Reactivity: Human