Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Antibody type:Primary
- Antigen name:integrin subunit beta 2
- Antigen symbol:ITGB2
- Clonality:Recombinant
- Clone:1B4
- Conjugation:Unconjugated
- ELISA:Yes
- Flow cytometry:Yes
- ImmunoFluorescence:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG kappa
- Reactivity:Rhesus monkey,Human
- Cross adsorption:No
- Form:Liquid
- Gene ID:3689
- Antigen synonyms:IB4|mIB4|Cell surface adhesion glycoproteins LFA-1/CR3/p150.95 subunit beta|ITGB2|Integrin beta-2|MFI7|m1B4|Complement receptor C3 subunit beta
- Modification:Unmodified
- Storage buffer:PBS with 0,02% Proclin 300
- Storage temperature:Store at 4 °C for up to 3 months. For longer storage, aliquot and store at −20 °C.
- Shipping temperature:Shipped on blue ice
- Immunogen:Blood was obtained from 4 helathy donors and the mononuclear cells harvested and grown. A BALB/c mouse was injected with 10^6 monocytes three times at monthly intervals, and four times with 5x10^6 monocytes oon each of the four days before fusion. The spleen cells were then fused to P3U1 myeloma cells to generate hybridomas and RIA was used to identify lines of interest
- Tested applications:RIA
- Pack type:Vial
- Pk:0,2 mg
Specifications
About this item
Type: Primary
Antigen: integrin subunit beta 2
Clonality: Recombinant
Clone: 1B4
Conjugation: Unconjugated
Epitope:
Host:
Isotype: IgG kappa
Reactivity: Human;Rhesus monkey