Order Entry
Northern Ireland
ContactUsLinkComponent
Anti-INSR Recombinant Antibody [Clone: 83-14]
Anti-INSR Recombinant Antibody [Clone: 83-14]
Catalog # PRSI24-174
Supplier:  ProSci Inc.
Anti-INSR Recombinant Antibody [Clone: 83-14]
Catalog # PRSI24-174
Supplier:  ProSci Inc.

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    insulin receptor
  • Antigen symbol:
    INSR
  • Clonality:
    Recombinant
  • Clone:
    83-14
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • ImmunoPrecipitation:
    Yes
  • Isotype:
    IgG kappa
  • Reactivity:
    Pig,
    Sheep,
    Human,
    Bovine
  • Western blot:
    Yes
  • Cross adsorption:
    No
  • Form:
    Liquid
  • Gene ID:
    3643
  • Antigen synonyms:
    IR|HHF5|Insulin Receptor alpha|CD220|HIR
  • Modification:
    Unmodified
  • Storage buffer:
    PBS with 0,02% Proclin 300
  • Storage temperature:
    Store at 4 °C for up to 3 months. For longer storage, aliquot and store at −20 °C.
  • Shipping temperature:
    Shipped on blue ice
  • Immunogen:
    This antibody was raised by immunizing mice with IM-9 lymphocytes followed by purified insulin receptor
  • Pack type:
    Vial
  • Pk:
    0,2 mg

Specifications

About this item

Type: Primary
Antigen: insulin receptor
Clonality: Recombinant
Clone: 83-14
Conjugation: Unconjugated
Epitope:
Host:
Isotype: IgG kappa
Reactivity: Human;Bovine;Pig;Sheep