Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Antibody type:Primary
- Antigen name:V-set immunoregulatory receptor
- Antigen symbol:Vsir
- Clonality:Recombinant
- Clone:13F3
- Conjugation:Unconjugated
- ELISA:Yes
- Flow cytometry:Yes
- Isotype:IgG lambda
- Cross adsorption:No
- Form:Liquid
- Gene ID:74048
- Antigen synonyms:Platelet receptor Gi24|V-domain Ig suppressor of T cell activation|Vsir|V-set immunoregulatory receptor|V-type immunoglobulin domain-containing suppressor of T-cell activation|PD-1H|Dies1|V-set domain-containing immunoregulatory receptor
- Modification:Unmodified
- Storage buffer:PBS with 0,02% Proclin 300
- Storage temperature:Store at 4 °C for up to 3 months. For longer storage, aliquot and store at −20 °C.
- Shipping temperature:Shipped on blue ice
- Immunogen:This antibody was raised by immunising Armenian hamsters with EL4 cells overexpressing VISTA-RFP, and VISTA-Ig fusion protein
- Tested applications:Blocking
- Pack type:Vial
- Pk:0,2 mg
Specifications
About this item
Type: Primary
Antigen: V-set immunoregulatory receptor
Clonality: Recombinant
Clone: 13F3
Conjugation: Unconjugated
Host: Recombinant
Isotype: IgG lambda
Reactivity: Mouse