Order Entry
Northern Ireland
ContactUsLinkComponent
Anti-Ptprc Recombinant Antibody [Clone: RA3-6B2]
Anti-Ptprc Recombinant Antibody [Clone: RA3-6B2]
Catalog # PRSI24-597
Supplier:  ProSci Inc.
Anti-Ptprc Recombinant Antibody [Clone: RA3-6B2]
Catalog # PRSI24-597
Supplier:  ProSci Inc.

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    protein tyrosine phosphatase, receptor type, C
  • Antigen symbol:
    Ptprc
  • Clonality:
    Recombinant
  • Clone:
    RA3-6B2
  • Conjugation:
    Unconjugated
  • Flow cytometry:
    Yes
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG kappa
  • Reactivity:
    Mouse
  • Cross adsorption:
    No
  • Form:
    Liquid
  • Gene ID:
    19264
  • Antigen synonyms:
    Lyt-4|C|Cluster designation 45R|B220|T200 glycoprotein|CD45|lymphocyte antigen 5|Ly-5|receptor type|T200 leukocyte common antigen|Leukocyte common antigen|T200|L-CA|lymphocyte common antigen|c polypeptide|protein tyrosine phosphatase|CD45 antigen|Receptor-type tyrosine-protein phosphatase C
  • Modification:
    Unmodified
  • Storage buffer:
    PBS with 0,02% Proclin 300
  • Storage temperature:
    Store at 4 °C for up to 3 months. For longer storage, aliquot and store at −20 °C.
  • Shipping temperature:
    Shipped on blue ice
  • Immunogen:
    This antibody was raised in Lou-M rats using a popliteal lymph node immunisation and fusion protocol as described by Martiniello et al (1994
  • Pack type:
    Vial
  • Pk:
    0,2 mg

Specifications

About this item

Type: Primary
Antigen: protein tyrosine phosphatase, receptor type, C
Clonality: Recombinant
Clone: RA3-6B2
Conjugation: Unconjugated
Host: Recombinant
Isotype: IgG kappa
Reactivity: Mouse