Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Antibody type:Primary
- Antigen name:serpin family A member 6
- Antigen symbol:SERPINA6
- Clonality:Recombinant
- Clone:SAIC-42B-10
- Conjugation:Unconjugated
- Isotype:IgG kappa
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Form:Liquid
- Gene ID:866
- Antigen synonyms:Corticosteroid-binding globulin|CBG
- Modification:Unmodified
- Storage buffer:PBS with 0,02% Proclin 300
- Storage temperature:Store at 4 °C for up to 3 months. For longer storage, aliquot and store at −20 °C.
- Shipping temperature:Shipped on blue ice
- Immunogen:Peptide 'AQLLQGLGFNLTER' derived from the Corticosteroid-binding globulin SERPINA6, conjugated to KL
- Tested applications:immunoMRM
- Pack type:Vial
- Pk:0,2 mg
Specifications
About this item
Type: Primary
Antigen: serpin family A member 6
Clonality: Recombinant
Clone: SAIC-42B-10
Conjugation: Unconjugated
Host: Recombinant
Isotype: IgG kappa
Reactivity: Human