Order Entry
Northern Ireland
ContactUsLinkComponent
Phenol red 0,4% in aqueous solution
Phenol red 0,4% in aqueous solution
Catalog # SGRO0673B
Supplier:  SOUTHERN GROUP
CAS Number:  
Phenol red 0,4% in aqueous solution
Catalog # SGRO0673B
Supplier:  SOUTHERN GROUP
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    100 mL
  • Seq:
    2440
  • Environmentally Preferable:

Specifications

About this item

Synonyms: 3,3-Bis(4-hydroxyphenyl)-3H-benzo[c][1,2]oxathiole 1,1-dioxide
 :
 :354.38 g/mol
EINECS:205-609-7
CAS Number:143-74-8
Formula:C₁₉H₁₄O₅S
MDL Number:MFCD00003552
Merck Index:13,07329