


Vinyl acetate stabilised for synthesis, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:108-05-4
:
Formula:C₄H₆O₂
MDL Number:MFCD00008713
:86.09 g/mol
Merck Index:12,10130
Boiling Pt:72,3 °C (1013 hPa)
Melting Pt:–100 °C
Density:0,932 g/cm³ (20 °C)
Storage Temperature:Ambient
UN:1301
ADR:3,II
:M-Clarity™ quality level = MQ200
EINECS:203-545-4
:GHS02
:GHS08
:GHS07
Specification Test Results
- Solubility:20 g/l (20 °C)
- Melting Point:-93 °C
- Molar Mass:86.09 g/mol
- Boiling Point:71 - 73 °C (1013 hPa)
- Vapor Pressure:120 hPa (20 °C)
- Flash Point:-8 °C
- Refractive Index:1.3954 (20 °C, 589 nm)
- Explosion Limit:2.6 - 13.4 %(V)
- Density:0.93 g/cm3 (20 °C)
- pH:7 (20 g/l, H2O, 20 °C)
- Ignition Point:385 °C
Specification Test Results
Frequently Bought Together