5-Nitro-8-quinolinol
Catalog # MOLE25352425-25G
Supplier: Molekula
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:25 g
- Seq:3308
Specifications
About this item
:
:190.16 g/mol
Merck Index:12,06753
Boiling Pt:419 °C (1013 hPa)
Melting Pt:179…181 °C
Density:1,549 g/cm³ (22 °C)
EINECS:223-662-4
CAS Number:4008-48-4
Formula:C₉H₆N₂O₃
MDL Number:MFCD00006791