
Patton-Reeders reagent (Calconcarboxylic acid) for complexometry, Fluka™
:3737-95-9
:
:C₂₁H₁₄N₂O₇S
:MFCD00004078
:438.42 g/mol
:300 °C
:223-117-0
:GHS07
:Ambient
- Identity (IR):complying
- Appearance of the solution:complying
- Water (Karl Fischer):Max. 7 %
- Suitability for complexometric Ca titration:complying
- Absorption maximum:638 - 648 nm
- Absorbtion (1%/1cm):Min. 100
Frequently Bought Together