Bromothymol blue sodium salt 0,04% (w/v) in aqueous solution
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:34722-90-2
:
Formula:C₂₇H₂₇Br₂NaO₅S
MDL Number:MFCD00077263
:646.37 g/mol
Storage Temperature:Ambient
EINECS:252-169-7
Related Information
Frequently Bought Together