Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
£0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Northern Ireland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Methyl red hydrochloride
Methyl red hydrochloride ACS
Methyl red hydrochloride ACS
:
Thermo Fisher Scientific
Methyl red hydrochloride ACS
:
Thermo Fisher Scientific
:
63451-28-5
:
:
C₁₅H₁₆ClN₃O₂
:
MFCD00012614
:
305.76 g/mol
:
175 °C (dec.)
:
264-190-9
:
GHS08
:
Ambient
msds_ALFA036668
Thermo Scientific Acroseal 25 Years
Frequently Bought Together
Dichloromethane ≥98% stabilised, TECHNICAL
: VWR Chemicals
Tetrahydrofuran ≥99% stabilised, GPR RECTAPUR®
: VWR Chemicals
Methanol, HiPerSolv CHROMANORM®, Ultra Plus for LC-MS, suitable for UPLC/UHPLC-MS instruments
: VWR Chemicals
Acetonitrile, LiChrosolv®, hypergrade for LC-MS, Supelco®
: Merck
Water, HiPerSolv CHROMANORM® for HPLC
: VWR Chemicals
Water, TECHNICAL, demineralised
: VWR Chemicals
Acetone ≥99.8%, HiPerSolv CHROMANORM® for HPLC
: VWR Chemicals
n-Hexane ≥95%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
: VWR Chemicals
Toluene ≥99%, GPR RECTAPUR® for pathology
: VWR Chemicals
Acetonitrile ≥99.5%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
: VWR Chemicals
2-Propanol ≥99.0%, GPR RECTAPUR®
: VWR Chemicals
Ethanol absolute ≥99.5%, GPR RECTAPUR®
: VWR Chemicals
n-Hexane ≥95%, TECHNICAL
: VWR Chemicals
Ethanol absolute, SupraSolv® for GC-ECD/FID
: Merck
Ethanol absolute ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
: VWR Chemicals
Methanol ≥99.9%, EMSURE® Reag. Ph. Eur., ACS, ISO for analysis, Supelco®
: Merck
Glycerine 85% Ph. Eur.
: VWR Chemicals
2-Methylbutane ≥99%, GPR RECTAPUR®
: VWR Chemicals
Tetrahydrofuran ≥99.9% unstabilised, HiPerSolv CHROMANORM® for LC-MS
: VWR Chemicals
Methanol ≥99.7%, PESTINORM® for pesticide residue analysis
: VWR Chemicals