Methyl red Reag. Ph. Eur. acid-base indicator, Fluka™
Supplier: Honeywell Chemicals
Certificates
About this item
Synonyms: 2-(4-Dimethylaminophenylazo)benzoic acid, 2-((4-(Dimethylamino)phenyl)diazenyl)benzoic acid
CAS Number:493-52-7
:
Formula:C₁₅H₁₅N₃O₂
MDL Number:MFCD00002425
Merck Index:12,06199
Melting Pt:179…182 °C
Density:0,989 g/cm³
:269.3 g/mol
Storage Temperature:Ambient
EINECS:207-776-1
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Indicator solution:complying
- Test of sensitivity:complying
- Change range (pH 4.4-6.0):complying
- UV spectrum:complying
Specification Test Results
Frequently Bought Together