Order Entry
Northern Ireland
ContactUsLinkComponent
4-Fluorostyrene 97% stabilised
Irritant
Flammable
4-Fluorostyrene 97% stabilised
Certificates
undefined
Irritant
Flammable
4-Fluorostyrene 97% stabilised

Specification Test Results

  • Appearance:
    Clear colorless to light yellow Liquid
  • Infrared spectrum:
    Conforms
  • Refractive index:
    1.5140 to 1.5160 (20°C, 589 nm)
  • GC:
    ≥96.0 %
  • Stabilizer:
    0.1 % tert.-butylcatechol

Specification Test Results

Related Information

Frequently Bought Together