

Chlorotoluron(3-(3-chloro-p-tolyl)-1,1-dimethylurea), extra pure
Catalog # 123702F
Supplier: VWR Chemicals
CAS Number:
Specifications
- Pack type:Glass ampoule
- Pk:10 mg
- Seq:2275
Specifications
About this item
:
:212.68 g/mol
Melting Pt:159…160 °C
Density:1,4 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:239-592-2
UN:3077
ADR:9,III
:GHS08
:GHS09
CAS Number:15545-48-9
Formula:(C₆H₃ClCH₃)NC(S)NH₂