Order Entry
Northern Ireland
ContactUsLinkComponent
Pinacidil monohydrate ≥98% (by HPLC)
Irritant
Pinacidil monohydrate ≥98% (by HPLC)
Catalog # ENZOALX550285M050
Supplier:  ENZO LIFE SCIENCES
CAS Number:  
undefined
Irritant
Pinacidil monohydrate ≥98% (by HPLC)
Catalog # ENZOALX550285M050
Supplier:  ENZO LIFE SCIENCES
CAS Number:  

Specifications

  • Pk:
    50 mg
  • Seq:
    2370

Specifications

Specification Test Results

  • Purity (HPLC):
    ≥98%

Specification Test Results

About this item

 :
 :263.3 g/mol
Merck Index:14,07438
 :GHS07
Storage Temperature:Ambient
CAS Number:85371-64-8
Formula:C₁₃H₁₉N₅·H₂O
MDL Number:MFCD00242745