Order Entry
Northern Ireland
ContactUsLinkComponent
 
Spectinomycin sulphate ≥98% (by HPLC)
Catalog # ENZOBMLA2810010
Supplier:  ENZO LIFE SCIENCES
CAS Number:  
Spectinomycin sulphate ≥98% (by HPLC)
Catalog # ENZOBMLA2810010
Supplier:  ENZO LIFE SCIENCES
CAS Number:  

Specifications

  • Pk:
    10 g
  • Seq:
    3236

Specifications

About this item

 :
 :430.43 g/mol
Storage Temperature:Freezer
CAS Number:23312-56-3
Formula:C₁₄H₂₄N₂O₇·H₂SO₄
MDL Number:MFCD00270185