Order Entry
Northern Ireland
ContactUsLinkComponent
 
ODN 1668 (synthetic), TLRgrade™, sterile, lyophilised
Catalog # ENZOALX746001C100
Supplier:  ENZO LIFE SCIENCES
CAS Number:  
ODN 1668 (synthetic), TLRgrade™, sterile, lyophilised
Catalog # ENZOALX746001C100
Supplier:  ENZO LIFE SCIENCES
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Pk:
    100 µG
  • Seq:
    1286

Specifications

Specification Test Results

  • Source:
    Synthetic
  • Purity :
    Activity and endotoxin tested - TLRgrade™
  • Endotoxin Content:
    <0.002 EU/µg
  • Formulation:
    Lyophilised, sterile

Specification Test Results

About this item

 :
 :6.383 g/mol
Storage Temperature:Refrigerator