Podophyllotoxin ≥98% (by HPLC)
Catalog # ENZOALX630086M100
Supplier: ENZO LIFE SCIENCES
CAS Number:
Specifications
- Pk:100 mg
- Seq:2440
Specifications
About this item
:
:414.41 g/mol
CAS Number:518-28-5
:4354-76-1
Formula:C₂₂H₂₂O₈
MDL Number:MFCD00075290