Order Entry
Northern Ireland
ContactUsLinkComponent
3,5-Dimethyl-1-adamantanamine hydrochloride ≥98% (by argentometric titration)
3,5-Dimethyl-1-adamantanamine hydrochloride ≥98% (by argentometric titration)
Catalog # ENZOALX550363M050
Supplier:  Enzo Life Sciences
CAS Number:  
undefined
3,5-Dimethyl-1-adamantanamine hydrochloride ≥98% (by argentometric titration)
Catalog # ENZOALX550363M050
Supplier:  Enzo Life Sciences
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Pk:
    50 mg
  • Seq:
    2370

Specifications

Specification Test Results

  • Purity (argentmetric titration):
    ≥98%

Specification Test Results

About this item

 :
 :215.77 g/mol
Merck Index:14,05829
Storage Temperature:Refrigerator
EINECS:255-219-6
CAS Number:41100-52-1
Formula:C₁₂H₂₁N
MDL Number:MFCD00214336