
Procainamide hydrochloride ≥98% (by HPLC)
Catalog # ENZOALX550287G010
Supplier: ENZO LIFE SCIENCES
CAS Number:
Specifications
- Pk:10 g
- Seq:3236
Specifications
Specification Test Results
- Purity (HPLC):≥98%
Specification Test Results
About this item
:
:271.789 g/mol
Merck Index:14,07756
EINECS:210-381-7
:GHS07
Storage Temperature:Ambient
CAS Number:614-39-1
Formula:C₁₃H₂₂ClN₃O
MDL Number:MFCD00012998