Order Entry
Northern Ireland
ContactUsLinkComponent
 
Lidocaine hydrochloride
Catalog # ENZOALX550251G005
Supplier:  ENZO LIFE SCIENCES
CAS Number:  
Lidocaine hydrochloride
Catalog # ENZOALX550251G005
Supplier:  ENZO LIFE SCIENCES
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

 :
EINECS:200-803-8
UN:2811
ADR:6.1,III
CAS Number:73-78-9
Formula:C₁₄H₂₃ClN₂O