Order Entry
Northern Ireland
ContactUsLinkComponent
5-(Diethylphosphono)-5-methyl-1-pyrroline-N-oxide ≥99% (by TLC)
5-(Diethylphosphono)-5-methyl-1-pyrroline-N-oxide ≥99% (by TLC)
Supplier:  Enzo Life Sciences
Certificates
undefined
5-(Diethylphosphono)-5-methyl-1-pyrroline-N-oxide ≥99% (by TLC)
Supplier:  Enzo Life Sciences

Specification Test Results

  • Purity (TLC):
    ≥99%
  • Appearance:
    Colorless to light yellow oil.

Specification Test Results

Frequently Bought Together