Resiquimod (synthetic) ≥98% (by HPLC), white to off-white solid endotoxin-free
Supplier: Enzo Life Sciences
Certificates
About this item
CAS Number:144875-48-9
:
Formula:C₁₇H₂₂N₄O₂
MDL Number:MFCD00937759
:314.389 g/mol
Storage Temperature:Freezer
Specification Test Results
- Purity:≥98% (HPLC)
- Identity:Determined by MS, ¹H-NMR and ¹³C-NMR
- Source:Synthetic
- Endotoxin Content:Endotoxin-free
Specification Test Results
Frequently Bought Together