
(+)-Catechin monohydrate ≥98% (by HPLC)
Catalog # ENZOALX385017G001
Supplier: ENZO LIFE SCIENCES
CAS Number:
Specifications
- Pk:1 g
- Seq:3047
Specifications
Specification Test Results
- Purity:≥98% (HPLC)
Specification Test Results
About this item
:
:290.3 g/mol (anhy)
Merck Index:14,01902
EINECS:205-825-1
:GHS07
Storage Temperature:Refrigerator
CAS Number:225937-10-0
:154-23-4
Formula:C₁₅H₁₄O₆·H₂O
MDL Number:MFCD00075649