Order Entry
Northern Ireland
ContactUsLinkComponent
Piericidin A (from Streptomyces sp. MST-AS5364) ≥95% (by HPLC)
Piericidin A (from Streptomyces sp. MST-AS5364) ≥95% (by HPLC)
Catalog # ENZOALX380235M002
Supplier:  Enzo Life Sciences
CAS Number:  
Piericidin A (from Streptomyces sp. MST-AS5364) ≥95% (by HPLC)
Catalog # ENZOALX380235M002
Supplier:  Enzo Life Sciences
CAS Number:  

Specifications

  • Pk:
    2 mg
  • Seq:
    2145

Specifications

About this item

 :
 :415.6 g/mol
Storage Temperature:Freezer
CAS Number:2738-64-9
Formula:C₂₅H₃₇NO₄
MDL Number:MFCD01677956