Kahweol ≥97% (by UPLC)
Catalog # ENZOALX350223M010
Supplier: Enzo Life Sciences
CAS Number:
Specifications
- Pk:10 mg
- Seq:2275
Specifications
Specification Test Results
- Purity:≥97% (UPLC)
- Source:Natural product isolated from the unsaponifiable fraction of petroleum ether extract of coffee beans
Specification Test Results
About this item
:
:314.2 g/mol
Storage Temperature:Freezer
CAS Number:6894-43-5
Formula:C₂₀H₂₆O₃