Cafestol ≥98% (by UHPLC)
ENZOALX350220M050
:
- Pk:50 mg
- Seq:2370
- Purity:≥98% (UHPLC)
- Identity:Determined by ¹H-NMR
- Source:Isolated from the unsaponifiable fraction of petroleum ether extract of coffee beans
:
:316.441 g/mol
:14,01634
:Freezer
:469-83-0
:C₂₀H₂₈O₃
:MFCD01075769