Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
£0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Northern Ireland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Aloesin
Aloesin ≥98% (by HPLC)
Aloesin ≥98% (by HPLC)
Catalog # ENZOALX350158M001
Supplier:
ENZO LIFE SCIENCES
CAS Number:
Expand image
Aloesin ≥98% (by HPLC)
Catalog # ENZOALX350158M001
Supplier:
ENZO LIFE SCIENCES
CAS Number:
Specifications
Pk:
1 mg
Seq:
2070
Specifications
About this item
Product Details
:
:
394.4 g/mol
CAS Number:
30861-27-9
Formula:
C₁₉H₂₂O₉
MDL Number:
MFCD00869744
Product Details