Order Entry
Northern Ireland
Orders LinkContactUsLinkComponent
Compound 15e ≥99% (by HPLC)

Compound 15e ≥99% (by HPLC)

5 mg
Catalog # ENZOALX270455M005
Supplier:   Enzo Life Sciences
Compound 15e ≥99% (by HPLC)
Catalog # ENZOALX270455M005
Supplier:  Enzo Life Sciences
CAS Number:  371943-05-4
Compound 15e ≥99% (by HPLC)

5 mg

CAS Number:  371943-05-4
Price
£340.00
Log in for availability
1 * 5 mg
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents

PI3 kinase inhibitor

Potent and specific inhibitor of phosphoinositide 3-kinasep110alpha isoform (IC50=0.0020µM). p110beta isoform (IC50=0.016µM)p110gamma isoform (IC50=0.66µM)C2beta isoform (IC50=0.22µM). Inhibits proliferation of A375 melanoma cells (IC50=0.58µM). Inhibits proliferation of U87MG human glioblastoma cells (IC50=1.10.µM).Inhibits proliferation of A2780 human ovarian cells (IC50=0.27µM).

Synonyms:  PI3K-α Inhibitor, 3-(4-Morpholinothieno[3,2-d]pyrimidin-2-yl)phenol
  • CAS Number:371943-05-4
  • Formula:C₁₆H₁₅N₃O₂S
  • Molecular Weight:313.4 g/mol
Specifications

Specifications

  • Pk:
    5 mg

Specifications

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment.