Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
£0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Northern Ireland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Ubiquinone-5
Ubiquinone-5 ≥95% (by HPLC)
Ubiquinone-5 ≥95% (by HPLC)
Ubiquinone-5 ≥95% (by HPLC)
Catalog # ENZOALX270294M002
Supplier:
ENZO LIFE SCIENCES
CAS Number:
Expand image
Ubiquinone-5 ≥95% (by HPLC)
Catalog # ENZOALX270294M002
Supplier:
ENZO LIFE SCIENCES
CAS Number:
Specifications
Pk:
2 mg
Seq:
2145
Specifications
About this item
Product Details
:
:
250.3 g/mol
CAS Number:
727-81-1
Formula:
C₁₄H₁₈O₄
Product Details