Order Entry
Northern Ireland
ContactUsLinkComponent
 
NUCLEAR-ID® ≥95% (by HPLC), CELLESTIAL® blue DNA stain
Catalog # ENZOENZ52406
Supplier:  ENZO LIFE SCIENCES
CAS Number:  
undefined
NUCLEAR-ID® ≥95% (by HPLC), CELLESTIAL® blue DNA stain
Catalog # ENZOENZ52406
Supplier:  ENZO LIFE SCIENCES
CAS Number:  

Specifications

  • Pk:
    200 µl
  • Seq:
    1358

Specifications

Specification Test Results

  • Excitation maximum::
    566nm
  • Emission maximum::
    650nm

Specification Test Results

About this item

 :
Storage Temperature:Freezer