Order Entry
Northern Ireland
ContactUsLinkComponent
Withaferin A (from Withania somnifera) ≥99% (by HPLC)
Withaferin A (from Withania somnifera) ≥99% (by HPLC)
Catalog # ENZOBMLCT1040010
Supplier:  ENZO LIFE SCIENCES
CAS Number:  
undefined
Withaferin A (from Withania somnifera) ≥99% (by HPLC)
Catalog # ENZOBMLCT1040010
Supplier:  ENZO LIFE SCIENCES
CAS Number:  

Specifications

  • Pk:
    10 mg
  • Seq:
    2275

Specifications

About this item

 :
 :470.6 g/mol
Melting Pt:241…243 °C
Storage Temperature:Freezer
CAS Number:5119-48-2
Formula:C₂₈H₃₈O₆