



Leukotriene A4 methyl ester ≥95% (by HPLC)
Supplier: Enzo Life Sciences
Certificates
About this item
CAS Number:73466-12-3
:
Formula:C₂₁H₃₂O₃
:332.5 g/mol
Storage Temperature:Ultra-Low Temperature Freezer (–80°C)
UN:3077
ADR:9,III
:GHS02
:GHS07
:GHS08
:GHS09
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specification Test Results
- Purity:≥95% (HPLC) (remainder 11-trans isomer)
- Formulation:Oil dissolved in 2% triethylamine/hexane.
- Concentration:1 mg/ml
- Source:Synthetic
Specification Test Results
Frequently Bought Together