Order Entry
Northern Ireland
ContactUsLinkComponent
Cytidine-5'-diphosphocholine sodium salt (CDP choline) ≥99%
Cytidine-5'-diphosphocholine sodium salt (CDP choline) ≥99%
Catalog # ENZOBMLEI1231000
Supplier:  ENZO LIFE SCIENCES
CAS Number:  
undefined
Cytidine-5'-diphosphocholine sodium salt (CDP choline) ≥99%
Catalog # ENZOBMLEI1231000
Supplier:  ENZO LIFE SCIENCES
CAS Number:  

Specifications

  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :510.31 g/mol
Storage Temperature:Freezer
CAS Number:33818-15-4
 :MFCD00150508
Formula:C₁₄H₂₆N₄NaO₁₁P₂
MDL Number:MFCD00037486